Assay Details
Gene Symbol: Hipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301304.1 | NP_001288233.1 | 4 - 5 | 1494 | 131 | |
NM_001301306.1 | NP_001288235.1 | 4 - 5 | 1604 | 131 | ||
NM_010432.2 | NP_034562.2 | 4 - 5 | 1604 | 131 | ||
NR_125731.1 | 4 - 5 | 1492 | 131 | |||
GenBank mRNA | AF071070.1 | - | 4 - 5 | 1494 | 131 | |
AF071071.1 | - | 4 - 5 | 1492 | 131 | ||
AF077658.1 | - | 4 - 5 | 1508 | 131 | ||
AF170303.1 | - | 4 - 5 | 1404 | 131 | ||
AF170304.1 | - | 4 - 5 | 1404 | 131 | ||
AK134408.1 | - | 4 - 5 | 1605 | 131 | ||
BC145697.1 | - | 4 - 5 | 1357 | 40131000 | 131 | |
BC145699.1 | - | 4 - 5 | 1357 | 40131002 | 131 | |
DQ279085.1 | - | 2 - 3 | 175 | 131 | ||
DQ279089.1 | - | 2 - 3 | 151 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
