Assay Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162500.1 | NP_001155972.1 | 3 - 4 | 323 | 65 | |
NM_022321.4 | NP_071716.3 | 3 - 4 | 278 | 65 | ||
GenBank mRNA | AK037365.1 | - | 3 - 4 | 250 | 65 | |
AK040914.1 | - | 3 - 4 | 209 | 65 | ||
AK079881.1 | - | 3 - 4 | 204 | 65 | ||
AK088419.1 | - | 3 - 4 | 228 | 65 | ||
BC011200.1 | - | 3 - 4 | 232 | 4190510 | 65 | |
BY200543.1 | - | 3 - 4 | 324 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
