Assay Details
Gene Symbol: Rps6kb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114334.1 | NP_001107806.1 | 8 - 9 | 883 | 75 | |
GenBank mRNA | AK012045.1 | - | 3 - 4 | 262 | 75 | |
AK032724.1 | - | 8 - 9 | 845 | 75 | ||
AK135716.1 | - | 8 - 9 | 851 | 75 | ||
AK163258.1 | - | 8 - 9 | 853 | 75 | ||
BC038491.1 | - | 8 - 9 | 856 | 4010710 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
