Assay Details
Gene Symbol: Zc3h7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081016.1 | NP_001074485.1 | 5 - 6 | 951 | 106 | |
GenBank mRNA | AK129268.1 | - | 5 - 6 | 628 | 106 | |
AK147484.1 | - | 5 - 6 | 715 | 106 | ||
AK147978.1 | - | 5 - 6 | 724 | 106 | ||
AK165492.1 | - | 5 - 6 | 743 | 106 | ||
AK171949.1 | - | 5 - 6 | 666 | 106 | ||
BC059923.1 | - | 5 - 6 | 706 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
