Assay Details
Gene Symbol: Zc3h7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081016.1 | NP_001074485.1 | 7 - 8 | 1087 | 77 | |
GenBank mRNA | AK129268.1 | - | 7 - 8 | 764 | 77 | |
AK147484.1 | - | 7 - 8 | 851 | 77 | ||
AK147978.1 | - | 7 - 8 | 860 | 77 | ||
AK165492.1 | - | 7 - 8 | 879 | 77 | ||
AK171949.1 | - | 7 - 8 | 802 | 77 | ||
BC059923.1 | - | 7 - 8 | 842 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
