Assay Details
Gene Symbol: Slc38a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175121.3 | NP_780330.2 | 2 - 3 | 497 | 74 | |
GenBank mRNA | AK017145.1 | - | 2 - 3 | 507 | 74 | |
AK018767.1 | - | 1 - 2 | 192 | 74 | ||
AK030917.1 | - | 2 - 3 | 508 | 74 | ||
AK031615.1 | - | 2 - 3 | 528 | 74 | ||
AK047403.1 | - | 2 - 3 | 531 | 74 | ||
AK129342.1 | - | 2 - 3 | 458 | 74 | ||
AK164568.1 | - | 1 - 2 | 80 | 74 | ||
BC027259.1 | - | 2 - 3 | 401 | 74 | ||
BC041108.1 | - | 2 - 3 | 367 | 4038546 | 74 | |
BC048178.1 | - | 2 - 3 | 497 | 6825801 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
