Assay Details
Gene Symbol: Ipo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153774.1 | NP_722469.1 | 6 - 7 | 756 | 96 | |
GenBank mRNA | AF273672.1 | - | 6 - 7 | 701 | 96 | |
AF273673.1 | - | 6 - 7 | 701 | 96 | ||
AJ309238.1 | - | 6 - 7 | 695 | 96 | ||
AK003047.1 | - | 4 - 5 | 96 | |||
AK036687.1 | - | 6 - 7 | 706 | 96 | ||
AK173127.1 | - | 6 - 7 | 696 | 96 | ||
BC056469.1 | - | 6 - 7 | 723 | 96 | ||
BC079546.1 | - | 7 - 8 | 867 | 96 | ||
BC098508.1 | - | 6 - 7 | 706 | 30015513 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
