Assay Details
Gene Symbol: Gucy1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204340.1 | NP_001191269.1 | 4 - 5 | 504 | 71 | |
NM_172810.3 | NP_766398.1 | 4 - 5 | 504 | 71 | ||
GenBank mRNA | AK047129.1 | - | 4 - 5 | 505 | 71 | |
AY236513.1 | - | 2 - 3 | 178 | 71 | ||
BC137868.1 | - | 4 - 5 | 501 | 8860887 | 71 | |
BC144988.1 | - | 4 - 5 | 501 | 9053536 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
