Assay Details
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009029.2 | NP_033055.2 | 15 - 16 | 1553 | 120 | |
GenBank mRNA | AK132875.1 | - | 4 - 5 | 299 | 120 | |
AK141028.1 | - | 15 - 16 | 1585 | 120 | ||
AK146467.1 | - | 10 - 11 | 884 | 120 | ||
BC096525.1 | - | 15 - 16 | 1553 | 6832808 | 120 | |
DQ400415.1 | - | 15 - 16 | 1403 | 120 | ||
M26391.1 | - | 15 - 16 | 1501 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
