Assay Details
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009029.2 | NP_033055.2 | 21 - 22 | 2338 | 122 | |
GenBank mRNA | AK132875.1 | - | 10 - 11 | 1084 | 122 | |
AK141028.1 | - | 21 - 22 | 2370 | 122 | ||
AK146467.1 | - | 16 - 17 | 1669 | 122 | ||
BC096525.1 | - | 21 - 22 | 2338 | 6832808 | 122 | |
DQ400415.1 | - | 21 - 22 | 2188 | 122 | ||
M26391.1 | - | 21 - 22 | 2286 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
