Assay Details
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177381.3 | NP_796355.2 | 18 - 19 | 2073 | 95 | |
GenBank mRNA | BC038030.1 | - | 19 - 20 | 2084 | 4020725 | 95 |
BC048005.1 | - | 14 - 15 | 1428 | 95 | ||
CO432313.1 | - | 5 - 6 | 509 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
