Assay Details
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177381.3 | NP_796355.2 | 19 - 20 | 2201 | 74 | |
GenBank mRNA | CF749874.1 | - | 1 - 2 | 86 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
