Assay Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146054.2 | NP_666166.2 | 11 - 12 | 1523 | 75 | |
GenBank mRNA | AK145030.1 | - | 11 - 12 | 1465 | 75 | |
AK166552.1 | - | 11 - 12 | 1462 | 75 | ||
BC033436.1 | - | 11 - 12 | 1444 | 4950810 | 75 | |
BC034168.1 | - | 2 - 3 | 177 | 75 | ||
CD550909.1 | - | 4 - 5 | 387 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC034168.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
