Assay Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146054.2 | NP_666166.2 | 13 - 14 | 1870 | 88 | |
GenBank mRNA | AK165608.1 | - | 13 - 14 | 1840 | 88 | |
AK166552.1 | - | 13 - 14 | 1809 | 88 | ||
BC033436.1 | - | 13 - 14 | 1791 | 4950810 | 88 | |
BC034168.1 | - | 4 - 5 | 524 | 88 | ||
CN716799.1 | - | 2 - 3 | 269 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
