Assay Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146054.2 | NP_666166.2 | 3 - 4 | 528 | 96 | |
GenBank mRNA | AK145030.1 | - | 3 - 4 | 470 | 96 | |
AK166552.1 | - | 3 - 4 | 467 | 96 | ||
BC033436.1 | - | 3 - 4 | 449 | 4950810 | 96 | |
BQ179963.1 | - | 2 - 3 | 328 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
