Assay Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146054.2 | NP_666166.2 | 6 - 7 | 1000 | 81 | |
GenBank mRNA | AK145030.1 | - | 6 - 7 | 942 | 81 | |
AK165608.1 | - | 6 - 7 | 970 | 81 | ||
AK166552.1 | - | 6 - 7 | 939 | 81 | ||
BC033436.1 | - | 6 - 7 | 921 | 4950810 | 81 | |
BU925369.1 | - | 3 - 4 | 364 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
