Assay Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146054.2 | NP_666166.2 | 7 - 8 | 1102 | 118 | |
GenBank mRNA | AK145030.1 | - | 7 - 8 | 1044 | 118 | |
AK165608.1 | - | 7 - 8 | 1072 | 118 | ||
AK166552.1 | - | 7 - 8 | 1041 | 118 | ||
BC033436.1 | - | 7 - 8 | 1023 | 4950810 | 118 | |
BU925369.1 | - | 4 - 5 | 466 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
