Assay Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146054.2 | NP_666166.2 | 8 - 9 | 1238 | 62 | |
GenBank mRNA | AK145030.1 | - | 8 - 9 | 1180 | 62 | |
AK165608.1 | - | 8 - 9 | 1208 | 62 | ||
AK166552.1 | - | 8 - 9 | 1177 | 62 | ||
BC033436.1 | - | 8 - 9 | 1159 | 4950810 | 62 | |
CD550909.1 | - | 1 - 2 | 102 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
