Assay Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146054.2 | NP_666166.2 | 9 - 10 | 1289 | 72 | |
GenBank mRNA | AK165608.1 | - | 9 - 10 | 1259 | 72 | |
AK166552.1 | - | 9 - 10 | 1228 | 72 | ||
BC033436.1 | - | 9 - 10 | 1210 | 4950810 | 72 | |
CD550909.1 | - | 2 - 3 | 153 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
