Assay Details
Gene Symbol: Smarca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290708.1 | NP_001277637.1 | 18 - 19 | 2569 | 138 | |
NM_053123.4 | NP_444353.3 | 18 - 19 | 2569 | 138 | ||
GenBank mRNA | AF325920.1 | - | 18 - 19 | 2576 | 138 | |
AK030741.1 | - | 18 - 19 | 2536 | 138 | ||
BC016071.1 | - | 18 - 19 | 2546 | 138 | ||
BC057115.1 | - | 18 - 19 | 2569 | 6406330 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
