Assay Details
Gene Symbol: Capn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130890.2 | NP_570960.2 | 18 - 19 | 1970 | 76 | |
GenBank mRNA | AB050202.1 | - | 18 - 19 | 1966 | 76 | |
AB061518.1 | - | 18 - 19 | 1916 | 76 | ||
BC125504.1 | - | 18 - 19 | 1942 | 40130019 | 76 | |
BC125508.1 | - | 18 - 19 | 1942 | 40130023 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
