Assay Details
Gene Symbol: Cenpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081363.2 | NP_001074832.2 | 1 - 2 | 107 | 74 | |
GenBank mRNA | AK080758.1 | - | 1 - 2 | 94 | 74 | |
AK165236.1 | - | 1 - 2 | 108 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK080758.1, AK165236.1, NM_001081363.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
