Assay Details
Gene Symbol: Cenpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081363.2 | NP_001074832.2 | 7 - 8 | 1189 | 148 | |
GenBank mRNA | AK029517.1 | - | 7 - 8 | 1140 | 148 | |
DQ642022.1 | - | 6 - 7 | 1046 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
