Assay Details
Gene Symbol: Cenpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081363.2 | NP_001074832.2 | 10 - 11 | 1565 | 86 | |
GenBank mRNA | AK029517.1 | - | 10 - 11 | 86 | ||
DQ642022.1 | - | 9 - 10 | 1422 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
