Assay Details
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145515.2 | NP_663490.2 | 13 - 14 | 1541 | 81 | |
GenBank mRNA | AB185282.1 | - | 13 - 14 | 1464 | 81 | |
AF453686.1 | - | 13 - 14 | 1471 | 81 | ||
AK173181.1 | - | 12 - 13 | 1391 | 81 | ||
BC112400.1 | - | 13 - 14 | 1470 | 40059725 | 81 | |
CX210969.1 | - | 5 - 6 | 601 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
