Assay Details
Gene Symbol: Ttll7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014905.1 | - | 10 - 11 | 1500 | 82 | |
AM690750.1 | - | 9 - 10 | 1141 | 82 | ||
AM690751.1 | - | 9 - 10 | 1141 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
