Assay Details
Gene Symbol: Ttll7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014905.1 | - | 12 - 13 | 1723 | 69 | |
AM690750.1 | - | 11 - 12 | 1364 | 69 | ||
AM690751.1 | - | 11 - 12 | 1364 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
