Assay Details
Gene Symbol: Ttll7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014905.1 | - | 14 - 15 | 1950 | 92 | |
AM690750.1 | - | 13 - 14 | 1591 | 92 | ||
AM690751.1 | - | 13 - 14 | 1591 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
