Assay Details
Gene Symbol: Ttll7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014905.1 | - | 1 - 2 | 192 | 79 | |
CJ110550.1 | - | 1 - 2 | 231 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK014905.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
