Assay Details
Gene Symbol: Ttll7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004213.1 | - | 1 - 2 | 72 | 77 | |
AM690750.1 | - | 3 - 4 | 279 | 77 | ||
AM690751.1 | - | 3 - 4 | 279 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
