Assay Details
Gene Symbol: Ttll7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014905.1 | - | 7 - 8 | 1077 | 82 | |
AM690750.1 | - | 6 - 7 | 718 | 82 | ||
AM690751.1 | - | 6 - 7 | 718 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
