Assay Details
Gene Symbol: Tmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177695.4 | NP_808363.3 | 3 - 4 | 475 | 62 | |
GenBank mRNA | AY236491.1 | - | 3 - 4 | 479 | 62 | |
AY263157.1 | - | 3 - 4 | 476 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
