Assay Details
Gene Symbol: Atad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099628.1 | NP_001093098.1 | 31 | 141 | ||
GenBank mRNA | AK003687.1 | - | 1 | 140 | ||
AK220160.1 | - | 7 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
