Assay Details
Gene Symbol: Pde8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008803.2 | NP_032829.1 | 12 - 13 | 1211 | 67 | |
GenBank mRNA | AF067806.1 | - | 1277 | 67 | ||
AK148238.1 | - | 1420 | 67 | |||
BC125578.1 | - | 12 - 13 | 1211 | 40130093 | 67 | |
BC132145.1 | - | 12 - 13 | 1211 | 40130422 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
