Assay Details
Gene Symbol: Bmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009755.3 | NP_033885.2 | 9 - 10 | 1420 | 83 | |
NR_033241.1 | 9 - 10 | 1457 | 83 | |||
GenBank mRNA | AK004995.2 | - | 9 - 10 | 1055 | 83 | |
AK147534.1 | - | 7 - 8 | 823 | 83 | ||
AK154239.1 | - | 9 - 10 | 1422 | 83 | ||
AK158218.1 | - | 9 - 10 | 1413 | 83 | ||
AK220295.1 | - | 9 - 10 | 1431 | 83 | ||
BC053923.1 | - | 9 - 10 | 1347 | 83 | ||
BC063079.1 | - | 5 - 6 | 550 | 83 | ||
BC066062.1 | - | 9 - 10 | 1348 | 6849066 | 83 | |
L24755.1 | - | 9 - 10 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
