Assay Details
Gene Symbol: Ncoa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010881.2 | NP_035011.1 | 6 - 7 | 1066 | 127 | |
GenBank mRNA | BC068177.1 | - | 3 - 4 | 374 | 30628581 | 127 |
BC080866.1 | - | 6 - 7 | 829 | 30532122 | 127 | |
U56920.1 | - | 5 - 6 | 624 | 127 | ||
U64606.1 | - | 3 - 4 | 369 | 127 | ||
U64828.1 | - | 6 - 7 | 979 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
