Assay Details
Gene Symbol: Dmpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190490.1 | NP_001177419.1 | 13 - 14 | 1677 | 98 | |
NM_032418.2 | NP_115794.1 | 12 - 13 | 1755 | 98 | ||
GenBank mRNA | BC075715.1 | - | 13 - 14 | 1585 | 4989259 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
