Assay Details
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008976.2 | NP_033002.2 | 3 - 4 | 876 | 80 | |
GenBank mRNA | AF170902.1 | - | 2 - 3 | 443 | 80 | |
AF170903.1 | - | 2 - 3 | 443 | 80 | ||
AK028783.1 | - | 3 - 4 | 840 | 80 | ||
AK037646.1 | - | 3 - 4 | 877 | 80 | ||
AK086524.1 | - | 3 - 4 | 870 | 80 | ||
AK156480.1 | - | 3 - 4 | 622 | 80 | ||
D31842.1 | - | 2 - 3 | 473 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
