Assay Details
Gene Symbol: Ntrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008746.5 | NP_032772.3 | 1 - 2 | 298 | 63 | |
NM_182809.2 | NP_877961.1 | 1 - 2 | 298 | 63 | ||
GenBank mRNA | AF035399.1 | - | 2 - 3 | 393 | 63 | |
AF035400.1 | - | 1 - 2 | 447 | 63 | ||
AY336094.1 | - | 1 - 2 | 243 | 63 | ||
BC139764.1 | - | 2 - 3 | 310 | 40110345 | 63 | |
JF417977.1 | - | 1 - 2 | 77 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
