Assay Details
Gene Symbol: Atp12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138652.2 | NP_619593.2 | 16 - 17 | 2541 | 100 | |
GenBank mRNA | BC109010.1 | - | 16 - 17 | 2539 | 100 | |
BC109011.1 | - | 16 - 17 | 2541 | 40048684 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
