Assay Details
Gene Symbol: Ahctf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026375.2 | NP_080651.2 | 2 - 3 | 789 | 60 | |
GenBank mRNA | AB059278.1 | - | 2 - 3 | 445 | 60 | |
BC138343.1 | - | 2 - 3 | 432 | 8861363 | 60 | |
BC138344.1 | - | 2 - 3 | 432 | 8861364 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
