Assay Details
Gene Symbol: Pcsk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291184.1 | NP_001278113.1 | 9 - 10 | 1312 | 96 | |
NM_011048.1 | NP_035178.1 | 9 - 10 | 1312 | 96 | ||
GenBank mRNA | D50060.1 | - | 9 - 10 | 1235 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
