Assay Details
Gene Symbol: 4930402F11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045940.1 | 1 - 2 | 74 | 64 | ||
GenBank mRNA | AK015048.1 | - | 1 - 2 | 76 | 64 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BY714574.1 | - | 76 | Chr. - - [-] on Build | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | arrestin domain containing 4 | Arrdc4 |
2410003C09Rik, AV216361 |
-746.9kb | Anti-sense | Chr. 7 - 68736994 - 68749238 on NCBI Build GRCm38 |
Downstream |
predicted gene, 29683 |
Gm29683 | 771.0kb | Anti-sense | Chr. 7 - 70267693 - 70339083 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2399878 | AK015048.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308909 | - | - |
Whole Mouse Genome G4122A | A_51_P427257 | AK015048.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205494 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205489 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205460 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05901784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205452 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205490 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205548 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205485 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06506402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205456 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205431 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205447 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205461 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205455 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06126517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06066116 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205479 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205484 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05901781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205487 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205502 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205449 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205491 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205480 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205440 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05901790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205500 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205450 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06506404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06032672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25205429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06126516 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1432402_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432402_at | - | - |
Mouse Genome 430 2.0 Array | 1432402_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700009380.mm.1 | TC0700001022.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700009380.mm.1 | NR_045940.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009380.mm.1 | NR_045940.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009382.mm.1 | TC0700001022.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700009382.mm.1 | NR_045940.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009382.mm.1 | NR_045940.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009385.mm.1 | TC0700001022.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700009385.mm.1 | NR_045940.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009385.mm.1 | NR_045940.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009383.mm.1 | TC0700001022.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700009383.mm.1 | NR_045940.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009383.mm.1 | NR_045940.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009384.mm.1 | TC0700001022.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700009384.mm.1 | NR_045940.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700009384.mm.1 | NR_045940.1 | Refseq |
Mouse Expression Set 430 | 1432402_PM_at | - | - |
Mouse Expression Set 430 | 1432402_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432402_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432402_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 649966 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 104730670 | - | - |
