Assay Details
Gene Symbol: Gm29856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK082514.1 | - | 2 - 3 | 463 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger and BTB domain containing 18 | Zbtb18 |
RP58, Zfp238, Znf238 |
-9.2kb | Sense | Chr. 1 - 177442356 - 177450764 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 2310043L19 gene | 2310043L19Rik | 180.9kb | Anti-sense | Chr. 1 - 177641542 - 177642943 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
