Assay Details
Gene Symbol: Traf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011633.2 | NP_035763.2 | 614 | 64 | ||
GenBank mRNA | AK141097.1 | - | 3 - 4 | 575 | 64 | |
AK144694.1 | - | 565 | 64 | |||
BC012702.1 | - | 568 | 4218625 | 64 | ||
D78141.1 | - | 591 | 64 | |||
D83528.1 | - | 456 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
