Assay Details
Gene Symbol: Parp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007415.2 | NP_031441.2 | 8 - 9 | 1224 | 77 | |
GenBank mRNA | AK028042.1 | - | 8 - 9 | 1243 | 77 | |
AK030889.1 | - | 8 - 9 | 1227 | 77 | ||
AK088679.1 | - | 8 - 9 | 1228 | 77 | ||
AK146677.1 | - | 8 - 9 | 1245 | 77 | ||
AK152360.1 | - | 8 - 9 | 1245 | 77 | ||
AK165158.1 | - | 8 - 9 | 1244 | 77 | ||
AK166104.1 | - | 8 - 9 | 1276 | 77 | ||
AK167699.1 | - | 8 - 9 | 1216 | 77 | ||
AK169326.1 | - | 8 - 9 | 1245 | 77 | ||
BC012041.1 | - | 8 - 9 | 1224 | 2648390 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
