Assay Details
Gene Symbol: Fh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010209.2 | NP_034339.2 | 8 - 9 | 1291 | 101 | |
GenBank mRNA | AK002379.1 | - | 8 - 9 | 1243 | 101 | |
AK147000.1 | - | 8 - 9 | 1285 | 101 | ||
BC002322.1 | - | 5 - 6 | 809 | 101 | ||
BC006048.1 | - | 8 - 9 | 3590691 | 101 | ||
BQ174129.1 | - | 2 - 3 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
