Assay Details
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009794.3 | NP_033924.2 | 13 - 14 | 1691 | 71 | |
GenBank mRNA | AF015038.1 | - | 13 - 14 | 1637 | 71 | |
AK052141.1 | - | 11 - 12 | 1279 | 71 | ||
AK150695.1 | - | 13 - 14 | 1665 | 71 | ||
AK152094.1 | - | 13 - 14 | 1665 | 71 | ||
AK152790.1 | - | 13 - 14 | 1668 | 71 | ||
AK152994.1 | - | 13 - 14 | 1668 | 71 | ||
AK154855.1 | - | 13 - 14 | 1705 | 71 | ||
AK183766.1 | - | 2 - 3 | 91 | 71 | ||
AK199295.1 | - | 2 - 3 | 71 | |||
BC054726.1 | - | 13 - 14 | 1691 | 5703563 | 71 | |
D38117.1 | - | 13 - 14 | 1732 | 71 | ||
Y10139.1 | - | 13 - 14 | 1573 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
