Assay Details
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010113.3 | NP_034243.2 | 1 - 2 | 509 | 99 | |
GenBank mRNA | AK084927.1 | - | 1 - 2 | 504 | 99 | |
AK085707.1 | - | 1 - 2 | 76 | 99 | ||
AK142666.1 | - | 1 - 2 | 205 | 99 | ||
BC060741.1 | - | 1 - 2 | 494 | 4164289 | 99 | |
BC092277.1 | - | 1 - 2 | 494 | 4240429 | 99 | |
J00380.1 | - | 1 - 2 | 504 | 99 | ||
V00741.1 | - | 1 - 2 | 460 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
