Assay Details
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281929.1 | NP_001268858.1 | 12 - 13 | 2327 | 89 | |
NM_001290451.1 | NP_001277380.1 | 12 - 13 | 2099 | 89 | ||
NM_016886.4 | NP_058582.3 | 12 - 13 | 2327 | 89 | ||
NR_104053.1 | 12 - 13 | 2327 | 89 | |||
GenBank mRNA | AB022342.1 | - | 12 - 13 | 2117 | 89 | |
AB079072.1 | - | 12 - 13 | 2063 | 89 | ||
AB102775.1 | - | 12 - 13 | 2063 | 89 | ||
AB102776.1 | - | 12 - 13 | 2063 | 89 | ||
AF483494.1 | - | 12 - 13 | 2063 | 89 | ||
AF483495.1 | - | 12 - 13 | 2063 | 89 | ||
AK135043.1 | - | 12 - 13 | 2081 | 89 | ||
AK138615.1 | - | 12 - 13 | 2232 | 89 | ||
AK220530.1 | - | 12 - 13 | 2114 | 89 | ||
BC076584.1 | - | 12 - 13 | 2189 | 89 | ||
BC086678.1 | - | 12 - 13 | 2272 | 30668476 | 89 | |
BC129855.1 | - | 12 - 13 | 2117 | 40109751 | 89 | |
BC129856.1 | - | 12 - 13 | 2117 | 40109752 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
